missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure

Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.

Supplier:  Thermo Scientific™ 21230010

Catalog No. 02-925-4



Explore available promotions

Description

Description

Collect, store and transport dry chemicals and environmental samples with Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure. Molded-in handgrips provide greater convenience and safety when pouring.

  • Useful for collecting, storing and transporting dry chemicals, environmental samples, reagents and distilled water
  • Good for freezing to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols
  • Square shape saves space, handles easily
  • Wide mouth gives easy access
  • Polypropylene cap included
  • Not liquid leakproof
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

Specifications

Specifications

4 L
Translucent
5.6 in.
89 mm
1
White
Yes
Wide
292 mm
5.66 in.
Polypropylene
142 mm
No
1 gal.
Large Wide-Mouth Square Bottle
3.5 in.
HDPE
Non-sterile
USP Class VI; FDA - 21 CFR
No
Square
Wide Mouth Bottle (Large)
11.5 in.
100 mm
Screw Cap
Videos
SDS
Documents

Documents

Promotions

Promotions