missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack

Carry out pharmaceutical and biotech applications with these LDPE dispensing tips for dropper bottles, which feature precise drip control.

Supplier:  Thermo Scientific™ 3127580001

 View more versions of this product

Catalog No. 01-814-233


Explore available promotions

Description

Description

Carry out pharmaceutical and biotech applications with Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles. Tips feature precise drip control and can be used with Nalgene LDPE Dropper bottles and Colored PP Closures.

Features:
  • Low density polyethylene
  • Offered in two drop sizes (40μL or 50μL)
  • Fits Nalgene Dropper Bottles Series 312750 and 312751
  • Use with Nalgene dropper closures for color coding your reagents

Ordering Information:

Dropper Bottles Dispensing Tips are bulk packed 2000 per case. Order Nalgene dropper bottles (Cat. Nos. 312750, 312751) and colored closure (Cat. No. 312760) separately.

Specifications

Specifications

50 μL
Clear
2000
Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle dispensing tips meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
40/50 μL
Dropping Bottle Tips
0.39 in.
11.22 mm
1.69 oz.
LDPE
2000
Yes
16.5 mm
Nalgene Dropper Closures
0.65 in.
Videos
SDS
Documents

Documents

Promotions

Promotions