Learn More
Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack
Carry out pharmaceutical and biotech applications with these LDPE dispensing tips for dropper bottles, which feature precise drip control.
Supplier: Thermo Scientific™ 3127590001
Description
Carry out pharmaceutical and biotech applications with Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles. Tips feature precise drip control and can be used with Nalgene LDPE Dropper bottles and Colored PP Closures.
Features:- Low density polyethylene
- Offered in two drop sizes (40μL or 50μL)
- Fits Nalgene Dropper Bottles Series 312750 and 312751
- Use with Nalgene dropper closures for color coding your reagents
Ordering Information:
Dropper Bottles Dispensing Tips are bulk packed 2000 per case. Order Nalgene dropper bottles (Cat. Nos. 312750, 312751) and colored closure (Cat. No. 312760) separately.
Specifications
| 40 μL | |
| Clear | |
| 2000 | |
| Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle dispensing tips meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| 40/50 μL | |
| Dropping Bottle Tips | |
| 0.39 in. | |
| 11.22 mm |
| 1.35 oz. | |
| LDPE | |
| 2000 | |
| Yes | |
| 16.5 mm | |
| Nalgene Controlled Dropper | |
| 0.65 in. |