Learn More
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles
Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
Supplier: Thermo Scientific™ 24010250
Description
High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
Features:- Polypropylene screw closure/stem with PPCO draw tube
- Tip can be cut back to provide increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
Specifications
8 oz., 250 mL | |
8 oz. | |
Stem and draw tube | |
36 | |
Yes | |
Narrow | |
175 mm | |
6.9 in. | |
24 mm | |
Screw Cap |
250 mL | |
Translucent | |
LDPE | |
6 | |
No | |
Cylindrical | |
Economy Wash Bottle | |
Polypropylene | |
61 mm | |
Yes |