Thermo Scientific™ Nalgene™ LDPE Sample Bags
Keep dry samples dry and moist samples moist with these LDPE sample bags, ideal for small lab items, botanical samples and other specimens.
Supplier:  Thermo Scientific™ 62550613
 View more versions of this product
Catalog No. 01-814-15C
Explore available promotions
Alert:
Not recommended for storing liquids.
Description
Description
Keep dry samples dry and moist samples moist with Thermo Scientific™ Nalgene™ LDPE Sample Bags. Ideal for small lab items, botanical samples and other specimens, these heavy-duty, double-track, low-density polyethylene zipper bags lock securely, but are not recommended for storing liquids.
  • Low-density polyethylene is clear for easy identification of contents
  • 4mil thick walls are strong to protect your samples
  • Bags are waterproof to keep contents dry
  • Transparent
Specifications
Specifications
Heavy-duty double-track zipper bag ideal for small lab items, botanical samples and other specimens
LDPE
33 cm
15.2 cm
Transparent
13 in.
6 in.