Thermo Scientific™ Nalgene™ LDPE Sample Vials with Closure
Store samples securely in these LDPE sample vials with closure, which have a friction-fit, snap closure and will not shatter like glass.
$47.33 - $138.50
Specifications
Material LDPE
No. per Case 144
No. per Pack 12
Product Line Nalgene
Closure Type Friction-fit Snap Closure
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Outer Height (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Outer Height (Metric) Price Quantity  
03-388A
View Documents
Thermo Scientific™
62500005
5 mL 23.6 mm 24.9 mm
Pack of 12 Each for $47.33
Case of 12 Pack for $567.96
03-388C
View Documents
Thermo Scientific™
62500012
12 mL 27.1 mm 35.6 mm
Pack of 12 Each for $47.93
Case of 12 Pack for $575.16
03-388E
View Documents
Thermo Scientific™
62500018
18 mL 27.1 mm 52.6 mm
Pack of 12 Each for $66.27
Case of 12 Pack for $795.24
03-388N
View Documents
Thermo Scientific™
62509028
28 mL 27.1 mm 84.6 mm
Pack of 12 Each for $99.77
Case of 12 Pack for $1,197.24
03-388P
View Documents
Thermo Scientific™
62509050
50 mL 37.3 mm 70.9 mm
Pack of 12 Each for $120.63
Case of 12 Pack for $1,447.56
03-388Q
View Documents
Thermo Scientific™
62509075
75 mL 37.3 mm 107.3 mm
Pack of 12 Each for $138.50
Case of 12 Pack for $1,662.00
Description
Description
Store samples securely in Thermo Scientific™ Nalgene™ LDPE Sample Vials with Closure, which have a friction-fit, snap closure and will not shatter like glass or polystyrene vials.
  • Straight sided vials allow easy access for filling and emptying
  • Friction fit closures are completely removable and snap in place for convenient vial access
  • Translucent wall allow user to see vials are filled or empty
  • Low density polyethylene is impact resistant and shatterproof even at temperatures down to -100°C
Specifications
Specifications
LDPE
12
Friction-fit Snap Closure
Certificate of Analysis (COA), Certificate of Conformance (COC)
144
Nalgene
144 Cases, 12 Pk.
Vial
SDS
missing translation for 'documents'
missing translation for 'documents'