Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack
Store, ship and package specialty chemicals, diagnostics, biologicals, reagents, adhesives and veterinary pharmaceuticals with these narrow-mouth HDPE bottles.
$518.35 - $2764.00
Specifications
Color | Translucent |
---|---|
Description | Narrow-Mouth Packaging Bottles |
Material | HDPE |
Closure Color | Natural, translucent |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
03-313-11A
|
Thermo Scientific™
3120890001 |
30 mL | 20-415 mm |
Case of 1000 Each for $2,484.71
|
|
||||
03-313-11B
|
Thermo Scientific™
3120890002 |
60 mL | 20-415 mm |
Case of 1000 Each for $2,764.00
|
|
||||
03-313-11C
|
Thermo Scientific™
3120890004 |
125 mL | 24-415 mm |
Case of 500 Each for $1,512.65
|
|
||||
03-313-11D
|
Thermo Scientific™
3120890008 |
250 mL | 24-415 mm |
Case of 250 Each for $1,151.97
|
|
||||
03-313-11E
|
Thermo Scientific™
3120890016 |
500 mL | 28-415 mm |
Case of 125 Each for $774.10
|
|
||||
03-313-11F
|
Thermo Scientific™
3120890032 |
1000 mL | 38-430 mm |
Case of 50 Each for $518.35
|
|
||||
Description
Safely store, ship and package liquid reagents using Thermo Scientific™ Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure, an ideal choice for specialty chemicals, diagnostics, biologicals, reagents, adhesives and veterinary pharmaceuticals. The injection blow-molded high-density polyethylene bottles feature a tough plastic construction with uniform wall thickness and strength. Polypropylene closures are included for leakproof† sealing.
Features:- Made from high-purity, food-grade resins
- Recommended for storage and shipment of liquid reagents
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
.Specifications
Translucent | |
HDPE | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
Narrow | |
Packer Bottle | |
Screw (Bulk Packed, Unassembled) |
Narrow-Mouth Packaging Bottles | |
Natural, translucent | |
Yes | |
Round | |
Polypropylene | |
No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.