missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles

Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.

Supplier:  Thermo Scientific™ 24250503

 View more versions of this product

Catalog No. 03-409-11C



Explore available promotions

Description

Description

Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.

Features:

  • Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
  • Self-venting, but contents stay in bottle with no risk of leakage
  • Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
  • Color bar that corresponds to colored closure to prevent cross-contamination
  • Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service
  • Tip that can be cut back to increase flow
  • Tight fit of draw tube and closure that prevents separation when squeezing bottle

Ordering Information:

Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.

Specifications

Specifications

16 oz., 500 mL
16 oz.
RTK Wash Bottle
24
Green
Yes
No
Cylindrical
Safety Wash Bottle
Methanol
28 mm
Screw Cap
500 mL
Natural
LDPE
6
Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)].
Not Autoclavable
Narrow
210.82 mm
8.3 in.
Polypropylene
72.64 mm
Yes
Videos
SDS
Documents

Documents

Promotions

Promotions