Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
Supplier: Thermo Scientific™ 24251002
Description
Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.
Features:
- Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
- Self-venting, but contents stay in bottle with no risk of leakage
- Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
- Color bar that corresponds to colored closure to prevent cross-contamination
- Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service
- Tip that can be cut back to increase flow
- Tight fit of draw tube and closure that prevents separation when squeezing bottle
Ordering Information:
Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.
Specifications
32 oz., 1000 mL | |
32 oz. | |
RTK Wash Bottle | |
12 | |
Natural | |
Yes | |
No | |
Cylindrical | |
Safety Wash Bottle | |
Ethyl Alcohol | |
38 mm | |
Screw Cap |
1000 mL | |
Natural | |
LDPE | |
4 | |
Meet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)]. | |
Not Autoclavable | |
Narrow | |
261.62 mm | |
10.3 in. | |
Polypropylene | |
91.44 mm | |
Yes |