Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.
Supplier:  Thermo Scientific™ 7121710530
 View more versions of this product
Catalog No. 02-923-16J
Explore available promotions
Description
Description
Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.
  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
Specifications
Specifications
Amber
Polypropylene
Yes
Round
Nalgene narrow-mouth and wide-mouth bottles
53 mm
Bottle Replacement Closures
12
Wide
Replacement Cap
Polypropylene
Videos
SDS
Documents
Documents
Promotions
Promotions
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.