Learn More
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle
Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles safe for food and beverage use
$18977.15 - $60462.82
Specifications
Closure Size | 20 mm |
---|---|
Color | Clear |
Diameter (Metric) Inner Neck | 11 mm |
For Use With (Application) | Sterile Sampling, Storage, and Shipment |
Material | PETG |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
14388030
|
Thermo Scientific™
CEN20350005 |
5 mL |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14388031
|
Thermo Scientific™
CEN20350020 |
20 mL |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Sterile to 10-6 SAL
- Non-pyrogenic and non-cytotoxic
- Leakproof High Density Polyethylene (HDPE) closures.
- Serum Vials are packed in shrink-wrapped trays with closures attached.
- Manufactured in compliance with ISO 13485:2003 quality requirements
Specifications
20 mm | |
11 mm | |
PETG | |
Not Autoclavable |
Clear | |
Sterile Sampling, Storage, and Shipment | |
ISO 13485:2003, 21CFR177.1520, USP Class VI |