missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).
$1699.57 - $2063.50
Specifications
Description | Serum Vials |
---|---|
Diameter (Metric) Inner | 11 mm |
Color | Clear |
Material | PETG |
Sterility | Sterile |
Catalog Number | Mfr. No. | Volume (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Closure Size | Price | Quantity | ||||
03-305-082
|
Thermo Scientific™
3420320005 |
5 mL | 20-415 |
Case of 1656 Each for $1,699.57
|
|
||||
03-313-801
|
Thermo Scientific™
3420320010 |
10 mL | 20-415 |
Case of 1260 for $2,063.50
|
|
||||
Description
Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).
- PETG
- Non-cytotoxic
- Non-pyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Sterile to 10-6 SAL
- Leakproof† with Nalgene lined continuous thread closures (Cat. No. 342158)
Ordering Information:
Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).
Specifications
Serum Vials | |
Clear | |
Sterile | |
Lined Continuous-thread Closure (Sold Separately) | |
Serum Vial | |
Lined Continuous-Thread Closure |
11 mm | |
PETG | |
Nalgene | |
ISO 13485:2016 | |
20-415 |