Learn More
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
$315.34 - $470.41
Specifications
| Material | PMP |
|---|---|
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Mouth Style | Narrow |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
|
10-042-5C
|
Thermo Scientific™
41090125 |
125 mL | 33 mm |
Pack of 6 Each for $378.31
Case of 4 Pack for $1,513.24
|
|
||||
|
10-042-5E
|
Thermo Scientific™
41090250 |
250 mL | 38 mm |
Pack of 4 Each for $323.66
Case of 3 Pack for $970.98
|
|
||||
|
10-042-5G
|
Thermo Scientific™
41090500 |
500 mL | 43 mm |
Pack of 4 Each for $470.41
Case of 3 Pack for $1,411.23
|
|
||||
|
10-042-5J
|
Thermo Scientific™
41091000 |
1000 mL | 53 mm |
Pack of 2 Each for $315.34
Case of 3 Pack for $946.02
|
|
||||
Description
Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.
Features:- Excellent chemical resistance to a wide variety of lab reagents
- More transparent than polypropylene but with similar chemical resistance
- Ideal for preparation and storage of culture media and titrations
- All sizes have easy-to-read graduations
- Autoclavable/Graduated
Includes:Polypropylene screw closures
.
Specifications
| PMP | |
| Reusable | |
| Narrow | |
| Plain |
| Autoclavable | |
| Yes | |
| Non-sterile | |
| Screw |