Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield
Simultaneously maintain safety and a clear field of vision by using this safety splash shield. Provides protection from splashes, aerosols and flying debris.
Supplier: Thermo Scientific™ 63560001
Alert:
Warning!: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.
Description
Simultaneously maintain safety and a clear field of vision by using the Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield. This shield attaches easily to benchtop shelving, machinery or other magnetic surfaces for convenient protection from splashes, aerosols and flying debris. This durable, scratch-resistant 1/8 in. (3mm) thick polycarbonate shield protects the user and features an easy to maneuver arm. Meets OSHA 29CFR1910.1030 standard.
- Flexible 14 inch arm makes it easy to maneuver shield over samples
- Magnetic base firmly grips to magnetic surfaces letting you position the shield horizontally (flat), vertically or on an overhead surface
- Provides UV protection from 200-360nm for added versatility
- Biohazard/Transparent
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
3 mm | |
21.5 cm | |
Meets OSHA 29CFR1910.1030 standard | |
Clear | |
12 in. | |
1 Ea. | |
Polycarbonate |
8.5 in. | |
0.12 in. | |
Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm | |
Safety Splash Shield | |
4/Cs. | |
Polycarbonate (Lens) | |
Yes |