missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Supplier: Thermo Scientific™ 42500075
Description
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
- Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
- Autoclavable
Specifications
76 mm | |
7 mm | |
Autoclavable | |
80 mm |
114 mL | |
Polypropylene | |
Funnel Analytical PP 75mm | |
6/Pk.; 36/Cs. |