missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Supplier: Thermo Scientific™ 42500100
Description
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
- Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
- Autoclavable
Specifications
98 mm | |
9 mm | |
Autoclavable | |
100 mm |
254 mL | |
Polypropylene | |
Funnel Analytical PP 100mm | |
4/Pk.; 24/Cs. |