Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Carboy
For Use With: For use with large batches of culture media, vaccine product production, bulk media and chemical storage; Autoclavable; Closure Material: Polypropylene; Color: White
$377.35 - $1,149.79
Specifications
Autoclavable | Autoclavable |
---|---|
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Polypropylene Carboy with Handles |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
02-960-20C
|
Thermo Scientific™
22500020 |
10 L |
N/A
|
|
|||||
02-960-20A
|
Thermo Scientific™
22500050 |
20 L |
N/A
|
|
|||||
02-960-20B
|
Thermo Scientific™
22500130 |
50 L |
N/A
|
|
|||||
Description
Choose Thermo Scientific™ Nalgene™ Autoclavable Carboys for large batches of culture media, vaccine product production, bulk media and chemical storage. These carboys feature a large mouth for easy filling/dispensing, and are made of natural polypropylene with white 83B polypropylene closure and TPE (thermoplastic elastomer) gasket.
Customization options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.
Key Features:
- Convenient shoulder handles allow easy carrying and pouring, even with gloved hands
- Available in 10, 20, and 50 L sizes for a variety of biopharmaceutical applications
- Graduated to contain in 1 gal. and 5 L increments for ease of use
- Nalgene Top Works Fluid Transfer Systems (Cat. No. 2135) permits convenient liquid aseptic transfer
- Autoclavable/Graduated/Leakproof†
Ordering Information:
- Additional closure options are Nalgene Quick Filling/Venting Closures (Cat. No. 2158) or Nalgene Filling/Venting Closures (Cat. No. 2162)
- Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149)
- Custom Tubing Sets are available-please refer to Thermo Scientific Custom Tubing Set Configurator at www.thermoscientific.com/tubingconfigurator.com
Specifications
Autoclavable | |
83B | |
Polypropylene Carboy with Handles | |
Handles | |
1 Ea. | |
Non-sterile |
Polypropylene | |
White | |
Media/Buffer Preparation â, Media/Buffer Storage, Storage, Transportation â | |
Polypropylene | |
Carboy with Handle |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.