missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Combinethese flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
$231.45 - $524.41
Specifications
Material | PPCO |
---|---|
Autoclavable | Autoclavable |
Disposable | Reusable |
Mouth Style | Narrow |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
10-041-10B
|
Thermo Scientific™
41020125 |
125 mL | Stopper No. 3 |
Pack of 12 Each for $524.41
Case of 2 Pack for $1,048.82
|
|
||||
10-041-10C
|
Thermo Scientific™
41020250 |
250 mL | Stopper No. 6 |
Pack of 6 Each for $331.03
Case of 2 Pack for $662.06
|
|
||||
10-041-10E
|
Thermo Scientific™
41021000 |
1000 mL | Stopper No. 8 |
Pack of 2 Each for $231.45
Case of 3 Pack for $694.35
|
|
||||
10-041-10F
|
Thermo Scientific™
41022000 |
2000 mL | Stopper No. 10 |
Pack of 2 Each for $288.59
Case of 3 Pack for $865.77
|
|
||||
Description
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
- Translucent wall makes liquid level easily visible
- 2000mL size has molded-in graduations
- Mouth sized to accept standard sized stoppers (stopper sold separately)
- Repeatedly autoclavable for long life and rigorous use
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
PPCO | |
Reusable | |
Non-sterile |
Autoclavable | |
Narrow | |
Plain |