Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle
For Use With: Ideal for the most extreme service conditions; Autoclavable; Closure Material: Polypropylene; Color: White
$620.74 - $947.68
Specifications
Autoclavable | Autoclavable |
---|---|
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Heavy-Duty Vacuum Carboy |
Description
Ideal for the most extreme service conditions, the Thermo Scientific™ Nalgene™ Polypropylene Heavy Duty Vacuum Carboy is useful as a vacuum trap and will hold a full vacuum up to eight hours. Constructed of thick-walled natural polypropylene to provide added strength, it is supplied with a white 83B PP closure with thermoplastic elastomer (TPE) gasket. Customization options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.
- Autoclavable when closure threads are disengaged and vessel is empty
- Not rated for pressure use
Ordering Information:
- For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
- Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
- For 1 to 5 L sizes, please refer to Nalgene Heavy Duty Vacuum Bottles (Cat. No. 2126)
Specifications
Autoclavable | |
83B | |
Heavy-Duty Vacuum Carboy | |
Handles | |
1 Ea. | |
Non-sterile |
Polypropylene | |
White | |
Media/Buffer Preparation â, Media/Buffer Storage, Storage, Transportation â | |
Polypropylene | |
Carboy with Handle |