missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle

For Use With: Store and transport solids, powders and liquids; Autoclavable; Closure Material: Polypropylene; Color: White

$436.91 - $569.09

Specifications

Autoclavable Autoclavable
Closure Material Polypropylene
Closure Size 100-415 mm
Color White
Description Autoclavable Wide-Mouth Carboy with Handles
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-961-65A
View Documents
Thermo Scientific™
22350020
10 L
Each for $436.91
Case of 6 Each for $2,621.46
 
02-961-65C
View Documents
Thermo Scientific™
22350050
20 L
Each for $569.09
Case of 4 Each for $2,276.36
 
Description

Description

Store and transport solids, powders and liquids using Thermo Scientific™ Nalgene™ Wide-Mouth Autoclavable Polypropylene Carboys with Handles. These durable natural polypropylene (PP) carboys feature a convenient wide-mouth opening to promote ease of cleaning and dispensing. The wide-shoulder handles enable easy carrying and pouring, even with gloved hands.

  • Graduated to contain in 1 gal. and 5 L increments for ease of use
  • Wide-mouth opening can accommodate overhead mixers
  • Autoclavable
  • Leakproof† white polypropylene 100 mm screw closures
Specifications

Specifications

Autoclavable
100-415 mm
Autoclavable Wide-Mouth Carboy with Handles
Graduated to contain in 5 L/1 gal. increments
Polypropylene
1 Ea.
Non-sterile
Polypropylene
White
Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Handles
Wide
Carboy with Handle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.