Thermo Scientific™ Nalgene™ Tefzel ETFE, Quick Action Spigot
For Use With: DS2327-series fluorinated carboys
Supplier: Thermo Scientific™ 64320010
Description
Employ Thermo Scientific™ Nalgene™ Quick-Action ETFE Tefzel™ Spigots as replacement spigots for Nalgene DS2327-series fluorinated carboys. They are chemically resistant to selected solvents and other chemicals. Body and plug are made of Tefzel Ethylene tetrafluoroethylene. Accepts tubing with 5/8 in. I.D.
Features:- Autoclavable (unassembled)
- Tubing adapter not included
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage of compatible solutions, but direct UV exposure should be avoided.
Specifications
Quick-Action Spigot ETFE fits 1-1/8 in. spigot boss for fluorinated carboys 1-1/8 inch | |
16 mm | |
ETFE | |
1 Ea. |
5/8 in. | |
DS2327-series fluorinated carboys | |
2/Cs. | |
Spigot |