Learn More
Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
For Use With: Ideal for use when visual inspection of contents is necessary; Autoclavable; Closure Material: Polypropylene; Color: Clear
Supplier: Thermo Scientific™ 23220050
Description
Easily fill and clean the Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Spigot, which features a wide-mouth opening, a sturdy stainless-steel handle and space-saving rectangular design. Clear construction allows easy viewing of carboy contents. Super tough; impact resistant. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features
- Molded-in graduations in 1 gallon and 5L increments for ease in dispensing
- Stainless-steel handle for ease in transporting containers
- Molded-in spigot boss provides strength and leak resistance
- A clear wall makes this carboy ideal when visual inspection of contents is necessary
- Follow autoclave instructions provided with the product for proper function; tubing adapter is not autoclavable
- 20L size has molded-in hand grips in the sidewall for easier handling
- Autoclavable/Transparent/Graduated
Includes:
Polypropylene spigot and screw closure
Specifications
Autoclavable | |
20 L | |
100 mm | |
Autoclavable Clear Rectangular Carboy w/Spigot | |
Ideal for use when visual inspection of contents is necessary | |
Molded-in spigot boss w/tubing adapter | |
Polycarbonate | |
1 Ea. | |
320 mm |
5.3 gal. | |
Polypropylene | |
Transparent | |
86 mm | |
399 mm | |
229 mm | |
4/Cs. | |
Carboy with Spigot |