Thermo Scientific™ Nalgene™ Reusable Filter Units
Eliminate the need for a manifold or breakable glass vacuum flask. Nalgene Reusable Filter Holders with Receiver are complete, environmentally-friendly systems.
$402.53 - $781.00
Catalog Number | Mfr. No. | Volume (Metric) Receiver | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) Receiver | Price | Quantity | |||||
09-740-23A
|
Thermo Scientific™
3004000 |
250 mL |
Each for $402.53
Case of 4 Each for $1,610.12
|
|
|||||
09-740-23D
|
Thermo Scientific™
3004050 |
500 mL |
Each for $600.09
Case of 4 Each for $2,400.36
|
|
|||||
09-740-23E
|
Thermo Scientific™
3004100 |
1000 mL |
Each for $781.00
Case of 4 Each for $3,124.00
|
|
|||||
Description
Reusable filter units include two membrane support plates: a clear one-piece plate for sterilization applications and a two-piece white plate for analytical applications. Accepts 47 or 50 mm diameter membranes. Two side-arms allow connection to vacuum line. Tubing adapters accept 1/4- to 5/16-inch (6 to 8 mm) I.D. vacuum tubing. 4 units per case.
Features- Replaces metal or breakable glass – major components are made of durable, transparent, autoclavable polysulfone
- Use for sterilization or analysis. Filter holders come with sterilization and analytical membrane support plates
- Designed to be used with vacuum or pressure
- Graduated chambers for easy reference
Specifications
Autoclavable | |
Polysulfone | |
1 |