missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ Carboy with Closure

For Use With: Large-volume media and culture preparation, particularly when visual inspection is required.; Autoclavable; Closure Material: Polypropylene; Color: Clear

$672.70 - $901.17

Specifications

Autoclavable Autoclavable
Closure Material Polypropylene
Closure Size 83B
Color Clear
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-962-6A
View Documents
Thermo Scientific™
22510020
10 L
Each for $672.70
Case of 4 Each for $2,690.80
Only null left
 
02-962-6B
View Documents
Thermo Scientific™
22510050
20 L
Each for $901.17
Case of 4 Each for $3,604.68
Only null left
 
Description

Description

Conduct large-volume media and culture preparation with these Nalgene™ Round Polycarbonate Clearboy™ Carboys, particularly when visual inspection is required. This polycarbonate Clearboy is durable, lighter and safer than glass, autoclavable for sterile applications and ideal for refrigerated or frozen storage of aqueous solutions such as sterile water, aqueous reagents and media.

Features:

  • Available in 10 and 20 L sizes for a variety of biopharmaceutical applications
  • Graduated in liters and gallons
  • Molded of transparent, durable polycarbonate
  • Customization options available for fluid transfer applications
  • Autoclavable, graduated, leakproof†

Includes:

  • Polypropylene closure and TPE gasket

Ordering Information:

Additional closure options include Nalgene Quick Filling/Venting Closures (Cat. No. 2158), Nalgene Filling/Venting Closures (Cat. No. 2162) Nalgene Top Works™ Fluid Transfer Systems (Cat. No. 2135) and Nalgene Validation Bottles (Cat. No. DS2127). Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149).

Note:

Completely disengage closure threads and disassemble spigot before autoclaving

Specifications

Specifications

Autoclavable
83B
Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
4/Cs.
Carboy
Non-sterile
Polypropylene
Clear
Polycarbonate
1 Ea.
Round
Upstream media/buffer prep and storage. Upstream process solution storage
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.