missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.

Supplier:  Thermo Scientific™ 41122800

 View more versions of this product

Catalog No. 03-600-150


Explore available promotions

Description

Description

Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features:
  • Sterile and single use: Reduce contamination risk
  • Collapses when autoclaved: Reduce bio hazardous waste
  • Graduated for quick volume assessment
Specifications

Specifications

95 oz.
PETG
1
None
Single-use
Narrow
Plain
70 mm
2800 mL
4
Erlenmeyer
Not Autoclavable
Yes
Sterile
Solid, non-vented
Videos
SDS
Documents

Documents

Promotions

Promotions

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.