missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure

Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.

Supplier:  Thermo Scientific™ 20180125

 View more versions of this product

Catalog No. 03-311-1Q



Explore available promotions

Description

Description

Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Square sides allow for good use of space and an easy grip
  • Caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Graduated for easy dispensing
  • Leakproof†
  • Can be used to -100°C (-148°F)
  • Not autoclavable
  • Participates in My Green Labs' ACT Label program

Includes:

Leakproof† Polypropylene Screw Caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided

Specifications

Specifications

4 oz., 125 mL
4 oz.
Narrow-Mouth Square Bottle
1.1 in.
Leakproof polypropylene screw closures
72
Non-sterile
Yes
Yes
Yes
Square
109 mm
2.12 in.
Polypropylene
53 mm
No
125 mL
Translucent
2.1 in.
28 mm
HDPE
12
Natural, translucent
USP Class VI; FDA - 21 CFR
Not Autoclavable
Narrow
Yes
Square Narrow Mouth Bottle
4.3 in.
38-430 mm
Screw Cap
Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.