Learn More
Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with these square bottles.
Supplier: Thermo Scientific™ 3420400250
Description
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure. These light-weight bottles are shatter-resistant, sterile and come in a space-saving square design.
Features:- PET with natural HDPE closure
- Space saving square design
- Molded in graduations and neck ring for use with tamper evident shrink sleeves
- Sterile, non-pyrogenic, non-cytotoxic
- Excellent gas barrier, with transparency and reduced permeability to CO2 and O2
- Bottle and closure material are ADCF (Animal Derived Component Free)
- Leakproof†
Recommended for: Storage and shipping of liquid media, buffers and sera.
Ordering Information:
Products are packaged in shrink-wrap trays. Trays are double polybagged inside the master carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
250 mL | |
Clear | |
1.1 in. | |
PET | |
30 | |
PET Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI, USP <661.1> | |
Yes | |
Square | |
140 mm | |
5.5 in. | |
59 mm |
8.5 oz. | |
2.3 in. | |
28 mm | |
60 | |
Sterile | |
Yes | |
Narrow | |
Sterile | |
Media Bottle | |
38-430 mm |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.