Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure, ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.
$927.74 - $1478.45
Specifications
| Color | Clear |
|---|---|
| For Use With (Application) | Media/Buffer Preparation â, Media/Buffer Storage, Storage |
| Material | PETG |
| Sterility | Sterile |
| Temperature Range (Metric) | -70°C to 70°C |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
03-311-1V
|
Thermo Scientific™
20190030 |
30 mL |
Case of 96 Each for $1,142.10
|
|
|||||
|
03-311-1W
|
Thermo Scientific™
20190060 |
60 mL |
Case of 96 Each for $1,478.45
|
|
|||||
|
03-312-1
|
Thermo Scientific™
20190125 |
125 mL |
Case of 48 Each for $1,031.23
|
|
|||||
|
03-312-3
|
Thermo Scientific™
20190250 |
250 mL |
Case of 48 Each for $1,286.26
|
|
|||||
|
03-312-4
|
Thermo Scientific™
20190500 |
500 mL |
Case of 24 Each for $927.74
|
|
|||||
|
03-312-5
|
Thermo Scientific™
20191000 |
1 L |
Case of 24 Each for $1,223.42
|
|
|||||
|
03-311-1X
|
Thermo Scientific™
20192000 |
2000 mL |
Case of 12 Each for $1,064.48
|
|
|||||
Description
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure. Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates, and also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
Features:
- Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
- Suitable for frozen storage down to -40°C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Sterile and non-pyrogenic
- Available in low particulate format
- Supported by extensive validation documentation
Ordering Information:
- Packed in shrink-wrapped trays
Note:
- 2 L size has molded-in handgrips
Warranty:
- 90 days
Specifications
| Clear | |
| PETG | |
| -70°C to 70°C | |
| Yes | |
| Narrow | |
| Media Bottle | |
| Not Autoclavable |
| Media/Buffer Preparation â, Media/Buffer Storage, Storage | |
| Sterile | |
| Bottles and closures are certified radiation sterilized (SAL 10-6), non-pyrogenic and non-cytotoxic, and comply with USP Class VI guidelines. | |
| Yes | |
| Square | |
| HDPE | |
| Upstream media/buffer prep and storage. Upstream process solution storage. Upstream Validation |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.