Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays
Package biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe in these durable, break-resistant media bottles.
$932.71 - $2085.32
Specifications
Color | Transparent |
---|---|
Description | With septum closure |
Material | PETG |
Sterility | Sterile |
Certifications/Compliance | Meet USP Class IV standards. Noncytotoxic. |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Fit for Purpose | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Fit for Purpose | Price | Quantity | |||
03-313-900
|
Thermo Scientific™
3420230060 |
60 mL | 24 mm | Sterile |
Case of 200 Each for $2,085.32
|
|
|||
03-313-901
|
Thermo Scientific™
3420230125 |
125 mL | 38-430 mm | Sterile |
Case of 96 Each for $1,480.19
|
|
|||
03-313-902
|
Thermo Scientific™
3420230500 |
500 mL | 38-430 mm | Sterile |
Case of 40 Each for $1,053.94
|
|
|||
03-313-903
|
Thermo Scientific™
3420231000 |
1 L | 38-430 mm | Sterile |
Case of 24 Each for $932.71
|
|
|||
14-178-159
|
Thermo Scientific™
3420239500 |
500 Expanded | 38-430 mm | Sterile |
Case of 48 Each for $1,138.91
|
|
|||
Description
Thermo Scientific™ Nalgene Square PETG Media Bottles are durable, break-resistant, and feature a septum closure for aseptic syringe dispensing or addition of supplements.
Features:- HDPE closure with silicone/PTFE septum provides a PTFE fluid contact surface that is inert and nonstick to ensure fluid integrity.
- Leakproof†, transparent media bottles
- Square design saves valuable shelf space
- Bottles and closures are sterile to 10-6 sterility assurance level
Ordering Information:
Packed in shrink-wrapped trays in double-polybagged cartons for cleanroom applications.
Recommended for:
Ideal for packaging of biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe.
Note:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Transparent | |
PETG | |
Meet USP Class IV standards. Noncytotoxic. | |
Yes | |
Square | |
Square Bottle |
With septum closure | |
Sterile | |
Yes | |
Wide | |
Sterile |