Learn More
Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles with Closure
Easily fill these square, wide-mouth HDPE bottles that offer excellent chemical resistance, making them suitable for a wide variety of uses.
$91.00 - $279.32
Specifications
Color | Translucent |
---|---|
Description | Wide-Mouth Square Bottle |
Material | HDPE |
Sterility | Non-sterile |
Closure Color | Natural, translucent |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
03-312AA
|
Thermo Scientific™
21140002 |
60 mL | 28 mm |
Pack of 12 Each for $91.00
Case of 6 Pack for $546.00
|
|
||||
03-312A
|
Thermo Scientific™
21140006 |
175 mL | 38 mm |
Pack of 12 Each for $146.00
Case of 6 Pack for $876.00
|
|
||||
03-312B
|
Thermo Scientific™
21140008 |
250 mL | 43 mm |
Pack of 12 Each for $195.97
Case of 6 Pack for $1,175.82
|
|
||||
03-312C
|
Thermo Scientific™
21140016 |
500 mL | 53 mm |
Pack of 12 Each for $279.32
Case of 4 Pack for $1,117.28
|
|
||||
03-312D
|
Thermo Scientific™
21140032 |
1 L | 63 mm |
Pack of 6 Each for $207.58
Case of 4 Pack for $830.32
|
|
||||
Description
Easily fill Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles. HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses. Good for freezer storage to -100°C (-148°F). This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
- Wide mouth is easy to fill
- Bottle design saves space in freezer
- HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses
- Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
- Leakproof† when used with Nalgene closures
- Bottle and Carton are readily recyclable in most communities
- Manufactured from non-toxic materials
- Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
.
Specifications
Translucent | |
HDPE | |
Natural, translucent | |
Yes | |
Wide | |
Yes | |
Unlined | |
Screw Cap |
Wide-Mouth Square Bottle | |
Non-sterile | |
USP Class VI; FDA - 21 CFR | |
No | |
Square | |
Square Wide Mouth Bottle | |
Polypropylene | |
No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.