missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.

Supplier:  Thermo Scientific™ 21100016

 View more versions of this product

Catalog No. 03-311-2D



Explore available promotions

Description

Description

Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Resistant to acids, alcohols and bases
  • Contact-clear bottle provides visible liquid level
  • Square design provides a large area for labeling
  • Linerless polypropylene closures provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable

Note:

Completely disengage threads or remove cap before autoclaving.

Specifications

Specifications

16 oz., 500 mL
16 oz.
Wide-Mouth Square Bottle
1.7 in.
PPCO
12
Natural, translucent
Yes
No
Square
Square Wide Mouth Bottle
5.7 in.
Polypropylene
74 mm
No
500 mL
Translucent
2.9 in.
43 mm
48
Non-sterile
USP Class VI; FDA - 21 CFR
Autoclavable
Wide
146 mm
2.97 in.
Unlined
53 mm
Screw Cap
Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.