Thermo Scientific™ Nalgene™ Storm Water Samplers and Mounting Kit
A convenient and affordable device for collecting quarterly first flush storm water grab samples in compliance with EPA sampling requirements.
Supplier:  Thermo Scientific™ 11601000
 View more versions of this product
Catalog No. 14-831-551
Explore available promotions
Description
Description
The Nalgene™ Storm Water Sampler is a convenient and affordable device for collecting quarterly storm water grab samples in compliance with EPA sampling requirements. No more standing in the rain waiting for water to flow or missed sampling events. The Storm Water Sampler can collect a full one liter grab sample of first flush storm water runoff through a storm water ditch, stream or storm grate outfall. Simply position the sampler in its protective mounting tube prior to a rain event, and leave it in place until after the storm.
  • UV-stabilized high-density polyethylene tube
  • 24″ steel stake
  • Includes stainless-steel worm clamp and lanyard, nylon rivets, polyester screen, wire hanger and carabiner-style snap
Click here to watch our storm water sampler instructional video.
Specifications
Specifications
Mounting Kit
Storm Water Sampler Mount Kit
Polyethylene
Meets EPA NPDES MSGP (Fed. Reg. FRL-6880-5 10/20/00 and most state grab sampling requirements