missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles

Dispense contents without tipping or shaking by using these LDPE wash bottles, designed with a wide mouth for faster, easier filling.

Supplier:  Thermo Scientific™ 24021000

 View more versions of this product

Catalog No. 03-409-10F

Explore available promotions

Description

Description

Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.

Features:
  • Bottle and tubulation are molded in one piece for uniform leakproof† service
  • Tip delivers a uniform, ultrafine stream without sputtering, but can also be cut back to increase flow
  • Leakproof†

Includes:

Polypropylene screw closure

Specifications

Specifications

32 oz., 1000 mL
32 oz.
LDPE bottle/tubulation
12
Yes
Wide
209.6 mm
8.25 in.
43 mm
Screw Cap
1000 mL
Translucent
LDPE
2
No
Cylindrical
Wash Bottle
Polypropylene
96.5 mm
No
Videos
SDS
Documents

Documents

Promotions

Promotions