Thermo Scientific™ Nalgene™ Unwire™ Half-Racks: Resmer™ Manufacturing Technology
Organize and store samples with space-efficient half-racks, ideal for use in incubators, refrigerators, freezers, under lab hoods and on benchtops.
$68.38 - $78.91
Specifications
Autoclavable | Autoclavable |
---|---|
Material | RESMER |
Shape | Rectangular |
Shelves | 3 Tiers |
Catalog Number | Mfr. No. | Array | For Use With (Equipment) | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Array | For Use With (Equipment) | Color | Price | Quantity | ||||
14-809-101
|
Thermo Scientific™
59720013 |
6 x 6 | 13mm Tubes | White |
Each for $68.38
Case of 8 Each for $547.04
|
|
||||
14-809-102
|
Thermo Scientific™
59720016 |
6 x 6 | 16mm Tubes | White |
Each for $71.88
Case of 8 Each for $575.04
|
|
||||
14-809-103
|
Thermo Scientific™
59720020 |
4 x 5 | 20mm Tubes | White |
Each for $78.91
Case of 8 Each for $631.28
|
|
||||
14-809-142
|
Thermo Scientific™
59720025 |
4 x 4 | 25mm Tubes | White |
Each for $76.03
Case of 8 Each for $608.24
|
|
||||
14-809-105
|
Thermo Scientific™
59720030 |
3 x 3 | 30mm Tubes | White |
Each for $74.12
Case of 8 Each for $592.96
|
|
||||
14-809-116
|
Thermo Scientific™
59720313 |
6 x 6 | 13mm Tubes | Blue |
Each for $68.38
Case of 8 Each for $547.04
|
|
||||
14-809-117
|
Thermo Scientific™
59720316 |
6 x 6 | 16mm Tubes | Blue |
Each for $71.88
Case of 8 Each for $575.04
|
|
||||
14-809-118
|
Thermo Scientific™
59720320 |
4 x 5 | 20mm Tubes | Blue |
Each for $74.12
Case of 8 Each for $592.96
|
|
||||
14-809-145
|
Thermo Scientific™
59720325 |
4 x 4 | 25mm Tubes | Blue |
Each for $76.03
Case of 8 Each for $608.24
|
|
||||
14-809-119
|
Thermo Scientific™
59720330 |
3 x 3 | 30mm Tubes | Blue |
Each for $74.12
Case of 8 Each for $592.96
|
|
||||
14-809-120
|
Thermo Scientific™
59720413 |
6 x 6 | 13mm Tubes | Green |
Each for $69.97
Case of 8 Each for $559.76
|
|
||||
14-809-122
|
Thermo Scientific™
59720416 |
6 x 6 | 16mm Tubes | Green |
Each for $71.88
Case of 8 Each for $575.04
|
|
||||
14-809-124
|
Thermo Scientific™
59720420 |
4 x 5 | 20mm Tubes | Green |
Each for $76.03
Case of 8 Each for $608.24
|
|
||||
14-809-125
|
Thermo Scientific™
59720430 |
3 x 3 | 30mm Tubes | Green |
Each for $74.12
Case of 8 Each for $592.96
|
|
||||
14-809-126
|
Thermo Scientific™
59720513 |
6 x 6 | 13mm Tubes | Red |
Each for $69.97
Case of 8 Each for $559.76
|
|
||||
14-809-127
|
Thermo Scientific™
59720516 |
6 x 6 | 16mm Tubes | Red |
Each for $71.88
Case of 8 Each for $575.04
|
|
||||
14-809-128
|
Thermo Scientific™
59720520 |
4 x 5 | 20mm Tubes | Red |
Each for $76.03
Case of 8 Each for $608.24
|
|
||||
14-809-129
|
Thermo Scientific™
59720530 |
3 x 3 | 30mm Tubes | Red |
Each for $74.12
Case of 8 Each for $592.96
|
|
||||
Description
Organize and store samples with space-efficient Thermo Scientific™ Nalgene™ Unwire™ Half-Racks, which have all the features of full-sized Unwire racks but occupy half the space. Ideal for use in incubators, refrigerators, freezers, under lab hoods and on benchtops, these racks fit easily in ice or water baths and will not float. They are molded in four bright colors to help keep samples organized.
- Constructed with Thermo Scientific™ Resmer™ Manufacturing Technology.
- Racks are repeatedly autoclavable.
- Smooth finish leaves fewer places for contaminants to lodge; no welds to break, no sharp edges.
- Endplates accept large labels.
- Rack withstands most lab chemicals and temperatures up to 121°C for a variety of lab procedures.
- 15mL conical tubes fit 20mm racks; 50mL conical fit 30mm racks.
- No assembly is required.
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Autoclavable | |
RESMER | |
Rectangular | |
3 Tiers |