Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.
$139.82 - $176.76
Specifications
Description RTK Wash Bottle
Material LDPE
Unitary Yes
Certifications/Compliance Meet OSHA requirements for workplace chemical container labeling.
Closure Included Yes
View More Specs
Products 8
Catalog Number Mfr. No. Capacity (Metric) Chemical Label Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Chemical Label Price Quantity  
03-409-20C
Accessories View Documents
Thermo Scientific™
24360503
500 mL Methanol
Pack of 4 Each for $176.76
Case of 6 Pack for $1,060.56
03-409-20D
Accessories View Documents
Thermo Scientific™
24360504
500 mL Isopropanol
Pack of 4 Each for $176.76
Case of 6 Pack for $1,060.56
03-409-20E
Accessories View Documents
Thermo Scientific™
24360505
500 mL Distilled Water
Pack of 4 Each for $166.00
Case of 6 Pack for $996.00
03-409-20B
Accessories View Documents
Thermo Scientific™
24360502
500 mL Ethyl Alcohol
Pack of 4 Each for $166.00
Case of 6 Pack for $996.00
03-409-20F
Accessories View Documents
Thermo Scientific™
24360506
500 mL Sodium Hypochlorite
Pack of 4 Each for $169.41
Case of 6 Pack for $1,016.46
03-409-20A
Accessories View Documents
Thermo Scientific™
24360501
500 mL Acetone
Pack of 4 Each for $166.00
Case of 6 Pack for $996.00
03-409-21E
View Documents
Thermo Scientific™
24361005
1000 mL Distilled Water
Pack of 2 Each for $139.82
Case of 6 Pack for $838.92
03-409-21A
View Documents
Thermo Scientific™
24361001
1000 mL Acetone
Pack of 2 Each for $147.06
Case of 6 Pack for $882.36
Description
Description
Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.
  • Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
  • Unique 38mm colored vented closure designed to prevent dripping and self-dispensing
  • Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles) with integrated vent, color coded for easy identification
  • Color-coded bar corresponds to closure, preventing closure mix-ups and cross-contamination
  • Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Tip can be cut back to provide increased flow
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Specifications
RTK Wash Bottle
Yes
Yes
No
Cylindrical
No
LDPE
Meet OSHA requirements for workplace chemical container labeling.
Not Autoclavable
Narrow
Wash Bottle
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'