Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth Opaque Amber HDPE Bottles with Closure: Bulk Pack
Store, ship and pack light-sensitive reagents in these wide-mouthed bottles that are leakproof when used with Nalgene closures.
$1567.90 - $4843.45
Specifications
Color | Opaque Amber |
---|---|
Description | Wide-Mouth Bottle |
Material | HDPE |
Sterility | Non-sterile |
Closure Color | Opaque Amber |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
02-923-78
|
Thermo Scientific™
3121060004 |
125 mL |
Case of 500 Each for $4,843.45
|
|
|||||
02-923-79
|
Thermo Scientific™
3121060008 |
250 mL |
Case of 250 Each for $3,389.48
|
|
|||||
02-923-76
|
Thermo Scientific™
3121060016 |
500 mL |
Case of 125 Each for $2,584.48
|
|
|||||
02-923-77
|
Thermo Scientific™
3121060032 |
1 L |
Case of 50 Each for $1,567.90
|
|
|||||
Description
Store, ship and pack light-sensitive reagents in Thermo ScientificTM NalgeneTM Wide-Mouth Opaque Amber HDPE Bottles with Closure.
Features:- Opaque amber polypropylene closures
- Uniform walls that resist splitting and puncturing
- Wide mouth for easy filling and dispensing
- Bulk packed for cost-efficiency
- Leakproof† when used with Nalgene closures
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
Specifications
Opaque Amber | |
HDPE | |
Opaque Amber | |
Wide | |
Wide Mouth Bottle | |
Screw (Bulk Packed, Unassembled) |
Wide-Mouth Bottle | |
Non-sterile | |
Yes | |
Round | |
Polypropylene | |
No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.