missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Benzoyl-L-argininamide Hydrochloride Monohydrate 98.0+%, TCI America™
$57.21 - $57.21
Chemical Identifiers
CAS | 4299-03-0 |
---|---|
Molecular Formula | C13H20ClN5O2 |
Molecular Weight (g/mol) | 313.786 |
MDL Number | MFCD00035003 |
InChI Key | PYZACNCNNFUUDO-PPHPATTJSA-N |
Synonym | s-n-1-amino-5-guanidino-1-oxopentan-2-yl benzamide hydrochloride, n-alpha-benzoyl-l-argininamide hydrochloride, 2s-5-carbamimidamido-2-phenylformamido pentanamide hydrochloride, bz-arg-nh hcl, bz-arg-nh2 hcl, benzoyl-l-arginine amide hydrochloride, n-benzoyl-l-arginine amide hydrochloride, benzoyl-arginine amide monohydrochloride monihydrate, nalpha-benzoyl-l-argininamidehydrochloridemonohydrate, 2s-5-diaminomethylidene amino-2-phenylformamido pentanamide hydrochloride |
PubChem CID | 22871352 |
IUPAC Name | N-[(2S)-1-amino-5-(diaminomethylideneamino)-1-oxopentan-2-yl]benzamide;hydrochloride |
SMILES | C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B01021G
|
TCI America
B01021G |
1 g |
Each for $57.21
|
|
|||||
Chemical Identifiers
4299-03-0 | |
313.786 | |
PYZACNCNNFUUDO-PPHPATTJSA-N | |
22871352 | |
C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)N.Cl |
C13H20ClN5O2 | |
MFCD00035003 | |
s-n-1-amino-5-guanidino-1-oxopentan-2-yl benzamide hydrochloride, n-alpha-benzoyl-l-argininamide hydrochloride, 2s-5-carbamimidamido-2-phenylformamido pentanamide hydrochloride, bz-arg-nh hcl, bz-arg-nh2 hcl, benzoyl-l-arginine amide hydrochloride, n-benzoyl-l-arginine amide hydrochloride, benzoyl-arginine amide monohydrochloride monihydrate, nalpha-benzoyl-l-argininamidehydrochloridemonohydrate, 2s-5-diaminomethylidene amino-2-phenylformamido pentanamide hydrochloride | |
N-[(2S)-1-amino-5-(diaminomethylideneamino)-1-oxopentan-2-yl]benzamide;hydrochloride |
Specifications
4299-03-0 | |
C13H20ClN5O2 | |
1 g | |
PYZACNCNNFUUDO-PPHPATTJSA-N | |
N-[(2S)-1-amino-5-(diaminomethylideneamino)-1-oxopentan-2-yl]benzamide;hydrochloride | |
22871352 | |
≥98.0% (T) | |
Nalpha-Benzoyl-L-argininamide Hydrochloride Monohydrate |
White-Yellow | |
MFCD00035003 | |
s-n-1-amino-5-guanidino-1-oxopentan-2-yl benzamide hydrochloride, n-alpha-benzoyl-l-argininamide hydrochloride, 2s-5-carbamimidamido-2-phenylformamido pentanamide hydrochloride, bz-arg-nh hcl, bz-arg-nh2 hcl, benzoyl-l-arginine amide hydrochloride, n-benzoyl-l-arginine amide hydrochloride, benzoyl-arginine amide monohydrochloride monihydrate, nalpha-benzoyl-l-argininamidehydrochloridemonohydrate, 2s-5-diaminomethylidene amino-2-phenylformamido pentanamide hydrochloride | |
C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)N.Cl | |
313.786 | |
313.79 | |
Crystalline Powder |
Safety and Handling
TSCA : No