missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
$255.67 - $255.67
Chemical Identifiers
CAS | 30189-36-7 |
---|---|
Molecular Formula | C20H33N3O8 |
Molecular Weight (g/mol) | 443.50 |
MDL Number | MFCD00057898 |
InChI Key | IQVLXQGNLCPZCL-UGPWUYPHNA-N |
Synonym | boc-lys boc-osu, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, a,n, n, n,n-di-boc-l-lysine hydroxysuccinimide ester, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys boc-osu calc. based on dry substance, c/n, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n |
PubChem CID | 13875534 |
IUPAC Name | (2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate |
SMILES | CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6206806
|
Thermo Scientific Chemicals
H6206806 |
5 g |
Each for $255.67
|
|
||||||
Description
Nalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
30189-36-7 | |
443.50 | |
IQVLXQGNLCPZCL-UGPWUYPHNA-N | |
13875534 | |
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O |
C20H33N3O8 | |
MFCD00057898 | |
boc-lys boc-osu, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, a,n, n, n,n-di-boc-l-lysine hydroxysuccinimide ester, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys boc-osu calc. based on dry substance, c/n, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n | |
(2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate |
Specifications
184°C | |
White | |
MFCD00057898 | |
boc-lys boc-osu, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, a,n, n, n,n-di-boc-l-lysine hydroxysuccinimide ester, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys boc-osu calc. based on dry substance, c/n, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n | |
IQVLXQGNLCPZCL-UGPWUYPHNA-N | |
(2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate | |
443.50 | |
443.49 | |
97% | |
Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester |
30189-36-7 | |
C20H33N3O8 | |
1559007 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O | |
5 g | |
13875534 | |
Moisture sensitive | |
Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only