missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(tert-Butoxycarbonyl)-L-arginine 4-Nitroanilide Hydrochloride 98.0+%, TCI America™
Supplier: TCI America B1497100MG
Specifications
| Nalpha-(tert-Butoxycarbonyl)-L-arginine 4-Nitroanilide Hydrochloride | |
| Yellow | |
| MFCD00143585 | |
| Nalpha-Boc-L-arginine 4-Nitroanilide Hydrochloride, Boc-Arg-pNA.HCl | |
| CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)NC1=CC=C(C=C1)[N+](=O)[O-].Cl | |
| 430.89 | |
| 430.89 | |
| Crystalline Powder |
| 99306-64-6 | |
| C17H27ClN6O5 | |
| 100 mg | |
| OUVONNGVXPOXGB-ZOWNYOTGSA-N | |
| tert-butyl N-[(2S)-5-(diaminomethylideneamino)-1-(4-nitroanilino)-1-oxopentan-2-yl]carbamate;hydrochloride | |
| 13502326 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 99306-64-6 | |
| 430.89 | |
| OUVONNGVXPOXGB-ZOWNYOTGSA-N | |
| 13502326 | |
| CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)NC1=CC=C(C=C1)[N+](=O)[O-].Cl |
| C17H27ClN6O5 | |
| MFCD00143585 | |
| Nalpha-Boc-L-arginine 4-Nitroanilide Hydrochloride, Boc-Arg-pNA.HCl | |
| tert-butyl N-[(2S)-5-(diaminomethylideneamino)-1-(4-nitroanilino)-1-oxopentan-2-yl]carbamate;hydrochloride |
Safety and Handling
TSCA : No