Learn More
Promega Nano-Glo™ Dual-Luciferase™ Reporter Assay System
The Nano-Glo™ Dual-Luciferase Reporter (NanoDLR) Assay System is a homogeneous reagent system that allows you to sequentially detect the activities of firefly (Photinus pyralis) luciferase and NanoLuc luciferase (Nluc) from a single sample.
Supplier: Promega N1610
Description
- Ultra-Sensitive Detection of Firefly and NanoLuc(R) Luciferase Activities in a Single Sample
- Greater sensitivity at low expression levels
- Choose either firefly or NanoLuc(R) luciferase as primary reporters
- Improved reagent stability brings more flexibility to assay design
Spécifications
| 10 mL | |
| -30°C to -10°C |