Learn More
Promega Nano-Glo HiBiT Lytic Detection System, 10ml DFS Item
Accurately quantify Any HiBiT Tagged Protein Expressed in Cells in an antibody free, plate based method.
Supplier:  Promega N3030
Catalog No. PRN3030
Only null left
Explore available promotions
Description
Description
  • Accurately Quantify Any HiBiT−Tagged Protein Expressed in Cells
  • Sensitive bioluminescent protein detection
  • Simple add−and−read assay—no antibodies required
  • Quantitate over 7 logs of linear dynamic range
Specifications
Specifications
10 mL
-30°C to -10°C
Product Suggestions
Product Suggestions