Promega Nano-Glo Live Cell Assay System
The Nano-Glo™ Live Cell Assay System is a single-addition, non-lytic detection reagent used to measure NanoBiT or NanoLuc luminescence from living cells.
Supplier: Promega N2011
Description
- Measures NanoLuc(R) Luminescence in Living Cells
- Substrate and buffer optimized to enhance stability
- Use to detect NanoBiT(R) protein complementation or NanoLuc(R) reporter activity
- Monitors luminescence at a single time point or continuously for up to 2 hours without compromising cell viability.
Specifications
-30°C to -10°C | |
NanoBiT™ protein complementation or NanoLuc™ reporter activity | |
Substrate and Buffer | |
100 Assays |