Promega Nano-Glo™ Luciferase Assay System DFS Item
The Nano-Glo™ Luciferase Assay System provides a simple, single-addition reagent that generates a glow-type signal in the presence of NanoLuc luciferase with a half-life of approximately 120 minutes in commonly used tissue culture media.
Supplier:  Promega N1120
 View more versions of this product
Catalog No. PRN1120
Explore available promotions
Description
Description
  • Add-Mix-Measure assay for NanoLuc(R) Luciferase
  • Bright NanoLuc(R) reporter enables use in applications where sensitivity is limited
  • Add-and-read simplicity allows scaling from bench to HTS
  • Extended bright light output optimized for batch and continuous processing
Specifications
Specifications
2 x 1mL Nano-Glo™ Luciferase assay Substrate, 100mL Nano-Glo™ Luciferase Assay Buffer, integral Lysis Buffer reagent
100 mL
Provides a single addition reagent that generates a glow-type signal in the presence of NanoLuc™ luciferase
Nano-Glo Luciferase Assay Buffer
−30 to −10°C