Learn More
Thermo Scientific™ NanoDrop Lite Plus Spectrophotometers
Quickly and easily assess sample concentration and purity ratios with the entry-level Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer. The instrument delivers accurate measurements in an easy-to-use, affordable package. Ideal for teaching and busy research labs.
$12910.70 - $22974.55
Specifications
Absorbance Range | 0.04 to 30 Abs (10 mm equivalent) |
---|---|
Certifications/Compliance | UL/CSA and CE |
Compatibility | Local Control Only |
Connections | USB 2.0 Port |
Depth (Metric) | 22 cm |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
NDLPLUSUSCA
|
Thermo Scientific™
NDLPLUSUSCAN |
NanoDrop Lite Plus Spectrophotometer (no printer) |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NDLPLUSPRUC
|
Thermo Scientific™
NDLPLUSPRUSCAN |
NanoDrop Lite Plus Spectrophotometer with Printer |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Quickly and easily assess sample concentration and purity ratios with the Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer, the entry-level complement to the full-featured Thermo Scientific™ NanoDrop™ One/OneC and NanoDrop™ Eight Spectrophotometers. While offering fewer features than our premium models, this instrument delivers fast, accurate measurements in an easy-to-use, affordable package.
The NanoDrop Lite Plus Spectrophotometer is a valuable teaching tool and is ideal for quality check measurements in busy research labs as well. Measure purified DNA, RNA, and protein concentration up to 30 Abs and calculate critical A260/A280 and A260/A230 purity ratios using trusted NanoDrop technology.
- Standard measurements
- Measures purified DNA, RNA, and protein concentration up to 30 Abs
- Up to 1,500 ng/μL dsDNA
- Up to 1,200 ng/μL RNA
- Up to 45 mg/mL BS
- Calculates critical A260/A280 and A260/A230 purity ratios
- Measures purified DNA, RNA, and protein concentration up to 30 Abs
- Fast, accurate measurements from only 1–2 μL—saves precious samples, no waste
- Sample-retention technology uses surface tension to hold the sample in place—no cuvettes or expensive consumables are needed
- Intuitive interface
- Built-in touchscreen operation
- No computer required
- Data storage and transfer
- Last 1,000 measurements saved locally; export data to a USB device
- Optional printer available for freezer-compatible labels (not included)
- Compact and portable—fits on any lab bench, ready for your next experiment
- Affordably priced—stretch tight budgets
- Academic teaching—educate students on molecular biology principles and experimental techniques on an industry-leading instrument
- Academic research labs—facilitate your research and publication of your novel research findings
- Biotech start-ups—accelerate the develop of new products
Specifications
0.04 to 30 Abs (10 mm equivalent) | |
Local Control Only | |
22 cm | |
LCD Touchscreen | |
Microvolume UV-Vis Spectrophotometer | |
Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification | |
Stainless Steel (303), Quartz Fiber | |
1 | |
≤ 1.8 nm (FWHM at Hg 254 nm) | |
One year | |
27 cm |
UL/CSA and CE | |
USB 2.0 Port | |
2048-element CMOS Linear Image Sensor | |
Xenon Flash | |
230 nm, 260 nm, 280 nm | |
22 cm | |
≤ 5 seconds | |
1.0 mm and 0.2 mm (auto adjust) | |
Standalone control with data export via USB Flash Drive | |
± 1 nm |