missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ NaOH, 1.0 M

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.

$1,513.00 - $3,940.63

Specifications

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Melting Point 0°C
Boiling Point 100°C
Color Clear
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A4782901
View Documents
Gibco™
A4782901
20 L N/A
 
A4782902
View Documents
Gibco™
A4782902
200 L N/A
 
Description

Description

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.

Features of Gibco NaOH solutions include:
• 0.1 M, 0.5 M, and 1 M concentrations
• Multi-compendial grade raw materials
• cGMP manufacturing to 21CFR820
• 20 L and 200 L ready-to-use single-use bioprocessing containers

Flexibility
Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.

Applications of Gibco NaOH solutions
• pH adjustment
• Facility/equipment sanitization
• Chromatography column cleaning, sanitization and storage

Specifications

Specifications

Sodium Hydroxide
0°C
Clear
≥12 to ≤14
Bioprocess Container
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
95 to 100%
1310-73-2
100°C
1.0 M
HNaO
Caustic soda, Acidity regulatory, Lye or Soda lye
[OH-].[Na+]
40.00
Compendial
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.