missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphazoline hydrochloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 179661000

Catalog No. AC179661000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Naphazoline hydrochloride
550-99-2
100.0
99%
C14H14N2·ClH
100g
naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon
DJDFFEBSKJCGHC-UHFFFAOYSA-N
2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
11079
246.74
99%
98.5
Authentic
Plastic bottle
MFCD00012554
15, 6453
Solubility in water: 170g/L (20°C).
C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl
246.74
CHEBI:7470
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Do not eat,drink or smoke when using this product.
Obtain special instructions before use.

GHS Signal Word: Danger

EINECSNumber : 208-989-2

RTECSNumber : NJ4375000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only