missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthacene 97.0+%, TCI America™

Supplier:  TCI America N0001100MG

 View more versions of this product

Catalog No. N0001100MG


Explore available promotions

Description

Description

Specifications

Specifications

Naphthacene
357°C
MFCD00003702
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1
228.294
CHEBI:32600
≥97.0% (HPLC)
92-24-0
C18H12
100 mg
IFLREYGFSNHWGE-UHFFFAOYSA-N
tetracene
7080
228.29
Crystalline Powder

Chemical Identifiers

92-24-0
228.294
IFLREYGFSNHWGE-UHFFFAOYSA-N
7080
tetracene
C18H12
MFCD00003702
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
CHEBI:32600
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1
Videos
Safety and Handling

Safety and Handling

RTECSNumber : QI7605000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions