missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene-1,4,5,8-tetracarboxylic Dianhydride 97.0+%, TCI America™

Supplier:  TCI America N036925G

 View more versions of this product

Catalog No. N036925G


Explore available promotions

Description

Description

Specifications

Specifications

Naphthalene-1,4,5,8-tetracarboxylic Dianhydride
C14H4O6
25 g
YTVNOVQHSGMMOV-UHFFFAOYSA-N
268.18
268.18
Crystalline Powder
81-30-1
MFCD00006915
1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride
C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O
6678
≥97.0% (HPLC)

Chemical Identifiers

81-30-1
268.18
YTVNOVQHSGMMOV-UHFFFAOYSA-N
6678
C14H4O6
MFCD00006915
1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride
C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O
Safety and Handling

Safety and Handling

RTECSNumber : QK3695000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions