missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene-1-sulfonic acid hydrate, 98%, Thermo Scientific Chemicals

$127.63 - $441.33

Chemical Identifiers

CAS 6036-48-2
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003987
InChI Key PSZYNBSKGUBXEH-UHFFFAOYSA-N
Synonym naphthalene-1-sulfonic acid dihydrate, 1-naphthalenesulfonic acid dihydrate, naphthalene-1-sulfonic acid hydrate
PubChem CID 71431272
IUPAC Name naphthalene-1-sulfonic acid;dihydrate
SMILES OS(=O)(=O)C1=C2C=CC=CC2=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2178209
View Documents
Thermo Scientific Chemicals
B2178209
10 g
Each for $127.63
 
AAB2178218
View Documents
Thermo Scientific Chemicals
B2178218
50 g
Each for $441.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6036-48-2
208.23
PSZYNBSKGUBXEH-UHFFFAOYSA-N
71431272
OS(=O)(=O)C1=C2C=CC=CC2=CC=C1
C10H8O3S
MFCD00003987
naphthalene-1-sulfonic acid dihydrate, 1-naphthalenesulfonic acid dihydrate, naphthalene-1-sulfonic acid hydrate
naphthalene-1-sulfonic acid;dihydrate

Specifications

6036-48-2
C10H8O3S
10 g
14,6376
PSZYNBSKGUBXEH-UHFFFAOYSA-N
naphthalene-1-sulfonic acid;dihydrate
71431272
98%
89°C to 91°C (dried)
MFCD00003987
UN2585
naphthalene-1-sulfonic acid dihydrate, 1-naphthalenesulfonic acid dihydrate, naphthalene-1-sulfonic acid hydrate
OS(=O)(=O)C1=C2C=CC=CC2=CC=C1
208.23
208.23 (Anhydrous)
Naphthalene-1-sulfonic acid hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID

EINECSNumber : 201-610-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only