missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene-2,6-dicarboxylic acid, 98+%, Thermo Scientific Chemicals

$102.58 - $1403.42

Chemical Identifiers

CAS 1141-38-4
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.192
MDL Number MFCD00004105
InChI Key RXOHFPCZGPKIRD-UHFFFAOYSA-N
Synonym 2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711
PubChem CID 14357
ChEBI CHEBI:44460
IUPAC Name naphthalene-2,6-dicarboxylic acid
SMILES C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1686806
View Documents
Thermo Scientific Chemicals
A1686806
5 g
Each for $102.58
 
AAA1686814
View Documents
Thermo Scientific Chemicals
A1686814
25 g
Each for $391.95
 
AAA1686822
View Documents
Thermo Scientific Chemicals
A1686822
100 g
Each for $1,403.42
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1141-38-4
216.192
RXOHFPCZGPKIRD-UHFFFAOYSA-N
14357
naphthalene-2,6-dicarboxylic acid
C12H8O4
MFCD00004105
2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711
CHEBI:44460
C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O

Specifications

1141-38-4
Odorless
MFCD00004105
2051257
RXOHFPCZGPKIRD-UHFFFAOYSA-N
naphthalene-2,6-dicarboxylic acid
14357
216.19
Naphthalene-2,6-dicarboxylic acid
>300°C
C12H8O4
5 g
2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711
C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O
216.192
CHEBI:44460
≥98%
Safety and Handling

Safety and Handling

EINECSNumber : 214-527-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only